Would you like to log in again?
Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
6603-5 6603-11 6603-8 6603-3 6603-21 6603-4 6603-13 6603-9 6603-2 6603-10 6603-7 6603-12 6603-22 6603-6
76546-392EA 27.64 USD InStock
76546-392 76546-380 76546-390 76546-400 76546-378 76546-388 76546-376 76546-398 76546-386 76546-374 76546-396 76546-384 76546-394 76546-382
Precision Specific Gravity Hydrometers
Hydrometers
These precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
Each hydrometer has been calibrated for use at 60 °F.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty