Would you like to log in again?
Dual Scale Hydrometers, Specific Gravity/Baume
Supplier: Heathrow Scientific
6603DS-5 6604DS-04 6603DS-1 6603DS-1S 6603DS-7 6603DS-2 6603DS-2S 6603DS-13 6603DS-3 6603DS-6 6603DS-6S 6603DS-4
76546-408EA 26.44 USD InStock
76546-408 76546-418 76546-406 76546-416 76546-404 76546-436 76546-414 76546-402 76546-412 76546-422 76546-410 76546-420
Dual Scale Hydrometers, Specific Gravity/Baume
Hydrometers
Dual-Scale Hydrometers have two scales on the stem, covering different applications in one unit.
  • Calibrated for use at 60 °F
The Baume scale has two different scales: one for liquids heavier than water and one for liquids lighter than water.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty