Would you like to log in again?
Specific Gravity Hydrometers with Thermometer
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6603TS-1 6602TS-7 6602TS-8
76546-370EA 53.02 USD InStock
76546-370 76546-424 76546-368
Specific Gravity Hydrometers with Thermometer
Hydrometers
These specific gravity hydrometers feature an internal thermometer for calculating temperature corrections and are calibrated for use at 60 ℉.
  • The liquid fill of these internal thermometers is safety blue
The precision specific gravity hydrometer feature and internal thermometer for calculating temperature corrections.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty