Would you like to log in again?
Wide-Range Specific Gravity Hydrometers
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6604-1 6604-2 6604-5 6604-4 6604-3
76546-428EA 27.64 USD InStock
76546-428 76546-426 76546-434 76546-432 76546-430
Wide-Range Specific Gravity Hydrometers
Hydrometers
These wide range specific gravity hydrometers are ideal to use when you aren't sure of the approximate value of your liquid. These refractometers can also be used for measuring multiple liquids with different specific gravity amounts.
All of our specific gravity hydrometers are calibrated at 60 °F.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty