Would you like to log in again?
Battery Hydrometer and Syphon Set
Supplier: Heathrow Scientific
6605-5H 6605-OSY 6605-5
76546-444EA 40.3 USD InStock
76546-444 76546-442 76546-440
Battery Hydrometer and Syphon Set
Hydrometers
Our battery acid hydrometer set is used in many industrial settings including fleet maintenance, automotive services, solar-powered system maintenance, and general industrial applications.
  • The rubber parts are made of nitrile rubber, which is chemically inert and durable
  • Calibrated for use at 77 °F
A reinforced box provides storage and is compartmented to eliminate breakage.
Delivery information: The set includes a specific gravity hydrometer and glass syphon tube with a rubber bulb, nozzle, and clamp.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty