Would you like to log in again?
Baume Hydrometers
Supplier: Heathrow Scientific
6609-1 6609-10 6609-2 6609-3 6609-15 6609-9 6609-14 6609-4 6609-11 6609-8 6609-12 6609-5 6609-13
76546-448EA 27.64 USD InStock
76546-448 76546-458 76546-446 76546-468 76546-456 76546-466 76546-454 76546-464 76546-452 76546-462 76546-450 76546-460 76546-470
Baume Hydrometers
Hydrometers
VEE GEE scientific Baume scale hydrometers are for liquids heavier than water. The Baume scale is based on the following: Specific gravity = 145/(145 - degrees Baume).
  • Wide variety of Baume ranges available
Calibrated for use at 15.5 °C/60 °F.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty