Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium Chloride Hydrometers

Supplier: Heathrow Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

6611-2 6611-1
76546-474EA 27.64 USD InStock
76546-474 76546-472
Sodium Chloride Hydrometers
Hydrometers
These hydrometers measure the percentage of sodium chloride. Specifically, model 6611-1 measures the saturation percentage of sodium chloride in solutions and model 6611-2 measures the percentage by weight of sodium chloride in solutions.

  • 30.5 cm (12") length

Both models are calibrated for use at 15 °C (60 °F).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty