Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alcohol Tralle and Proof Hydrometers

Supplier: Heathrow Scientific
6612-2TS 6612-1
76546-480EA 54.81 USD InStock
76546-480 76546-478
Alcohol Tralle and Proof Hydrometers
Hydrometers
Our alcohol hydrometers measure using the tralle and proof scales.

  • SafetyBLUE fill
  • Thermometer (model 6612-2TS only) measures from −10 to +100 °F
  • Tralle/proof range 0 to 100%/0 to 200
  • Tralle/proof subdivisions/tolerance 1%/ ±1%, 2%/ ±2%
  • Calibration for use at 15.5 °C (60 °F)
  • Not for use with beer or wine

Verify the alcohol content of distilled spirits with our alcohol hydrometer with Tralle and proof scales. Model 6612-2TS includes an internal thermometer for measuring solution temperature.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty