Would you like to log in again?
IRS Alcohol Hydrometers
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6613-L 6613-Q 6613-H 6613-N 6613-K 6613-F 6613-G 6613-P 6613-R 6613-B 6613-I 6613-M
76546-518EA 59.12 USD InStock
76546-518 76546-506 76546-490 76546-526 76546-514 76546-502 76546-522 76546-510 76546-498 76546-486 76546-494 76546-482
IRS Alcohol Hydrometers
Hydrometers
These IRS alcohol hydrometers use the proof scale and comply with IRS specifications.
  • Calibration for use at 15.5 °C/60 °F
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty