Would you like to log in again?
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6613-K-C 6613-R-C 6613-P-C 6613-M-C 6613-F-C 6613-I-C 6613-Q-C 6613-L-C 6613-G-C 6613-N-C 6613-H-C 6613-B-C
76546-528EA 371.82 USD InStock
76546-528 76546-516 76546-504 76546-524 76546-512 76546-500 76546-488 76546-520 76546-496 76546-484 76546-508 76546-492
Alcohol Hydrometers, Proof Scale, IRS Specification, with Certification
Hydrometers
These IRS alcohol hydrometers use the Proof scale and comply with IRS specifications.
  • Calibration for use at 15.5 °C/60 °F
  • NIST-Traceable certificate
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
Delivery information: Includes hydrometer, NIST-traceable certificate, and PVC case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty