Would you like to log in again?
Plato Hydrometers with Thermometer
Supplier: Heathrow Scientific
6614TS-7 6614TS-8 6614TS-5 6614TS-6
76546-536EA 53.09 USD InStock
76546-536 76546-534 76546-532 76546-530
Plato Hydrometers with Thermometer
Hydrometers
Our Plato hydrometers are calibrated to show the concentration of extract as a percentage by weight in brewery wort. For example, a 5° P reading indicates 5 g of extract per 100 g of wort.
  • Calibration for use at 20 °C/68 °F
  • Spirit thermometer liquid fill
Primarily used in the brewing industry, the Plato scale is carried out at 20 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty