Would you like to log in again?
ASTM Specific Gravity Hydrometers
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
67132H 67116H 67117H 67126H 67111H 67115H 67130H 67131H 67128H 67118H 67125H 67114H 67112H 67120H 67119H 67134H 67133H 67113H 67127H 67129H
76546-558EA 34.4 USD InStock
76546-558 76546-568 76546-556 76546-578 76546-566 76546-588 76546-576 76546-554 76546-564 76546-586 76546-552 76546-574 76546-584 76546-562 76546-572 76546-550 76546-560 76546-582 76546-570 76546-580
ASTM Specific Gravity Hydrometers
Hydrometers
Our ASTM specific gravity hydrometers are calibrated at 60 °F and manufactured to the specifications of the American Society for Testing and Materials (ASTM).
  • Calibrated for use at 15.5 °C/60 °F
Select the hydrometer that you need by matching the ASTM number shown. Specific Gravity, also known as relative density, is the ratio of the mass of a liquid to the mass of an equal volume of distilled water.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty