Would you like to log in again?
Soil Hydrometer, Soil Colloids, ASTM 152H
Supplier: Heathrow Scientific
67152H
76546-592EA 60.22 USD InStock
76546-592
Soil Hydrometer, Soil Colloids, ASTM 152H
Hydrometers
This hydrometer measures the amount of soil colloids in grams of soil samples in suspension.
  • Calibrated for use at 20 °C (68 °F)
This hydrometer is often used with a sedimentation cylinder for soil analysis.
Certifications: Manufactured to the specifications of the American Society for Testing and Materials (ASTM).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty