Would you like to log in again?
API Thermo-Hydrometers
Supplier: Heathrow Scientific
6752HTS 6756HTS 6755HTS 6754HTS 6753HTS 6757HTS
76546-604EA 53.85 USD InStock
76546-604 76546-602 76546-600 76546-598 76546-596 76546-594
API Thermo-Hydrometers
Hydrometers
Our API scale hydrometers measure the scale which was selected in 1921 by the American Petroleum Institute (API), the U.S. Bureau of Mines, and the National Institute of Standards and Technology (NIST) as the standard hydrometer scale for petroleum products.
  • Safety blue thermometer liquid fill
The thermometer scale (where applicable) for these thermo-hydrometers is located in the body. All instruments are suitable for certification for use as a primary reference with NIST traceability.
The API scale is based on the following formula: °API at 60 °F = 141.5/(specific gravity at 60 °F) - 131.5.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty