Would you like to log in again?
Armored Encapsulated Glass Thermometers
Supplier: Heathrow Scientific
80904E-A 80903E-A 80706E-A 80701E-A 80906E-A 80704E-A 80902E-A 80707E-A 80708E-A 80702E-A 80901E-A 80908E-A 80703E-A 80907E-A
76546-638EA 74.31 USD InStock
76546-638 76546-616 76546-626 76546-656 76546-666 76546-644 76546-622 76546-632 76546-610 76546-686 76546-674 76546-662 76546-650 76546-680
Armored Encapsulated Glass Thermometers
Thermometers Analog Thermometers
Our precision, serialized thermometers are encapsulated in FEP. The armor is constructed of nickel-plated brass.
  • Includes FEP encapsulation and nickel-plated brass armor
  • Feature safety blue fill against a white background for easy reading
  • Wide variety of temperature ranges available
These are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications. In the event of breakage the FEP coating contains the broken glass and liquid fill, preventing injury or contamination. The armor protects the thermometer from rough handling and shock during use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty