Would you like to log in again?
Serialized Encapsulated Glass Thermometers
Supplier: Heathrow Scientific
80902E 80908E 80703E 80707E 80907E 80903E 80706E 80701E 80904E 80906E 80702E 80901E 80708E 80704E
76546-648EA 56.78 USD InStock
76546-648 76546-636 76546-614 76546-624 76546-678 76546-654 76546-608 76546-620 76546-664 76546-642 76546-630 76546-684 76546-672 76546-660
Serialized Encapsulated Glass Thermometers
Thermometers Analog Thermometers
Our precision, serialized thermometers are encapsulated in FEP. These are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications requiring serialization.
  • Includes FEP encapsulation
  • Feature safety blue fill against a white background for easy reading
  • Wide variety of temperature ranges available
In the event of breakage the FEP coating contains the broken glass and liquid fill, preventing injury or contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty