Would you like to log in again?
Soil and Compost Dial Thermometers
Supplier: Heathrow Scientific
82200-48 82100-36 82200-36
76546-698EA 100.5 USD InStock
76546-698 76546-696 76546-690
Soil and Compost Dial Thermometers
Thermometers Analog Thermometers
Our dial thermometers are an excellent choice for many applications including soil and compost testing, food and beverage quality control, and asphalt/concrete testing.
  • Dial diameter 76.2 mm (3")
The dial thermometer indicates temperature through the metal stem with attached circular scale. It does don’t require batteries, it is rugged, and readings can be adjusted with the calibration screw on the back.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty