Would you like to log in again?
Divosan® Plus, Non-foaming Liquid Peroxyacetic Acid Sanitizer
Supplier: Diversey
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Divosan®
57571100 3177186 3747804
76547-730EA 164.36 USD InStock
76547-730 76547-690 76547-692
Divosan® Plus, Non-foaming Liquid Peroxyacetic Acid Sanitizer
Cleaning Agents Detergents and Surface Cleansers
Divosan® Plus is a peracetic acid based, broad spectrum sanitizer.
  • Completely non-foaming and suitable for clean-in-place (CIP) and spray applications
  • Rapidly decomposes after use
  • Sanitizes (reduces bacteria) in 1 minute, controls spoilage organisms; controlsmolds and yeast
  • Effective at temperatures as low as 40°F
  • Acidic pH, suitable under CO2 environments
  • 1.0 - 5.7 ounces per 5 gallons of water (88-500 ppm peracetic acid)
It is suitable for use in dairies, breweries, soft drink and all other food processing operations.
Divosan Plus is completely non-foaming and sanitizes effectively at temperatures as low as 40°F.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty