Would you like to log in again?
SCA Sentol®, Acid Cleaner
Supplier: Diversey
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SCA Sentol®
9676470
76547-722CT 1810.88 USD InStock
76547-722
SCA Sentol®, Acid Cleaner
Cleaning Agents Detergents and Surface Cleansers
SCA Sentol® cleaner is a phosphoric acid-based low foaming liquid with excellent penetration and cleaning abilities for use on pharmaceutical and biotechnology processing equipment.
  • Penetrates and dissolves mineral deposits quickly and thoroughly
  • Optimum blend of cleaning/wetting agents provide rapid penetration and vigorous cleaning action
  • Free-rinsing for a streak-free and bright finish
  • Safe on stainless steel, glass and copper, when used as directed
  • Suitable for all application, methods, spray, soak circulation or manual
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty