Would you like to log in again?
Liquid Brill Tak VC85, Liquid Caustic Detergent
Supplier: Diversey
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Brill Tak VC85
57727300 57727150 57727280 57727010
76547-746GL 258747.41 USD InStock
76547-746 76547-748 76547-744 76547-750
Liquid Brill Tak VC85, Liquid Caustic Detergent
Cleaning Agents Detergents and Surface Cleansers
Liquid Bril Tak® detergent is a blended liquid caustic formulation for spray, soak and circulation cleaning of equipment and pipelines in food and beverage plants where water is of moderate to high hardness.
  • Simple to use with no need for additional additives
  • Suitable for bulk shipment and automatic feeding
  • Highly chelated formula prevents scale formulation in hard water
  • Inhibits water hardness precipitation during rinsing
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty