Would you like to log in again?
Soft Care® Instant Hand Sanitizer AF
Supplier: Diversey
Soft Care
100961733
76547-798CT 175.95 USD InStock
76547-798
Soft Care® Instant Hand Sanitizer AF
Skincare Soaps and Skin Cleansers
Soft Care® Instant Hand Sanitizer AF is a benzalkonium chloride based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protected against irritation.
  • Alcohol-free formulation for use where alcohol-based hand sanitizers are a concern
  • Safe, quick and effective hand rub helps reduce the spread of germs
  • Skin softening emollients leave hands soft and with no sticky residues
  • Dye-free, reducing potential skin irritation and allergic reactions for users with sensitive skin
  • NSF E-3 certified for use in food handling establishments
Dye-free and alcohol-free.
It is a preferred option for those facilities who do not desire alcohol based products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty