Would you like to log in again?
All Purpose Virex® Disinfectant Cleaner
Supplier: Diversey
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CBD540533 CBD540557 CBD540540
76547-814CT 58.48 USD InStock
76547-814 76547-816 76547-818
All Purpose Virex® Disinfectant Cleaner
Cleaning Agents Disinfectants
A ready-to-use, quaternary-based, hospital-use disinfectant that provides excellent cleaning and deodorizing in one step.
  • Meets OSHA's BloodBorne pathogen standards for HBV and HIV
  • One-step quaternary disinfectants cleaner and deodorant to clean and disinfect hard surfaces in hospitals, nursing homes and other facilities
  • Provides broad spectrum kill of microorganisms including HBV, HIV-1, VRE, MRSA, GRSA, MRSE, VISA, PRSP, Herpes Simplex Types 1&2, Influenza Type A2, Adenovirus, Rotavirus and many more
  • Ready to use
  • Kills the organisms that cause odors and works as an odor counteractant to further eliminate odors not associated with bacteria
Disinfects in three minutes.
Bactericide, tuberculocide, virucide, fungicide.
Kills Norovirus, VRE and MRSA.
Meets bloodborne pathogen standards for decontaminating blood and body fluids.
Colorless with a lemon scent.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty