Would you like to log in again?
VWR® Cover Cleanroom Paper
Supplier: VWR
VWR®
11-80-COVER500 11-80-COVER
76547-896CS 251.75 USD InStock
76547-896 76547-898
VWR® Cover Cleanroom Paper
Paper
Cover-weight latex-free paper has been injected with a synthetic copolymer to substantially reduce particle generation.
  • Exceptional thickness and strength, with excellent heat resistance
  • High opacity and 'class 10 certified'
Ideal for use in printers and high speed photocopies.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty