Would you like to log in again?
MyBrix Handheld Refractometer, Mettler-Toledo
Supplier: Mettler-Toledo
30693201 30693200
76547-950EA 4563.3 USD InStock
76547-950 76595-302
MyBrix Handheld Refractometer, Mettler-Toledo
Refractometers Handheld Refractometers
With a full Brix range and ten integrated sugar-related scales, the MyBrix offers fast measurements of almost any food and beverage sample. A compact, robust, and weatherproof design makes this portable device ideal for field and on-site measurements.
  • Measurements within seconds
  • Fits in any pocket
  • Robust and durable
  • Instant and affordable sugar analysis
With a full Brix range and ten integrated sugar-related scales, the MyBrix offers fast measurements of almost any food and beverage sample. A compact, robust, and weatherproof design makes this portable device ideal for field and on-site measurements.
The MyBrix measures refractive index and sugar content instantly and precisely: Results are received within just two seconds in the desired scale.
The MyBrix pocket refractometer is compact, lightweight, and designed to fit comfortably in the hand. With a weatherproof design (IP65 rated) and the rubberized keypad, MyBrix is ideal for measurements in the field and near the production line.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty