Would you like to log in again?
Fixative Bouins, Statlab
Supplier: StatLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11750-1 11750-2 11750-.5 11750-32
76547-976CS 149.94 USD InStock
76547-976 76547-978 76547-972 76547-974
Fixative Bouins, Statlab
Stains Microscopy and staining reagents
Saturated aqueous picric acid/formalin solution.
  • Bouins is commonly used in trichrome staining procedures and with GI tract specimens
Packaging: Manufactured in the USA
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty