Would you like to log in again?
Fixative Rapid Differential Stain Hema-Diff™, Statlab
Supplier: StatLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11985-16 11985-1
76547-988EA 112.59 USD InStock
76547-988 76547-990
Fixative Rapid Differential Stain Hema-Diff™, Statlab
Stains Microscopy and staining reagents
The original StatLab best-selling rapid stain for the past 40 years. Three-step procedure for differential staining of blood smears, used extensively in FNA cytology.
  • Entire procedure takes 30 seconds as air dried specimens are fixed in Solution I, then dipped in Solutions II and III
  • Excellent for FNA cytology, differential staining of blood smears, microbiological organisms, H. pylori, and bone marrow specimens
  • Versatile and very economical
  • Components sold separately or as a kit
  • Can also be used as a special stain for Helicobacter pylori. 16 oz Hema-Diff #1 Fixative
Part of the StatLab Hema-Diff Rapid Differential Stain Line.
Manufactured in the USA
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty