Would you like to log in again?
Decal™ Decalcifier, StatLab
Supplier: StatLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1211-5 1211-1 1211-32
76547-994EA 28.76 USD InStock
76547-994 76547-996 76547-992
Decal™ Decalcifier, StatLab
Stains Microscopy and staining reagents
Since 1957 Original Decal® has been the industry standard. A gentle decalcifier formulated from semiconductor grade hydrochloric acid, EDTA, and highly purified reagent grade water. Can be used to safely decalcify all types of tissue samples from needle biopsies to cortical bone. It’s mild enough that most samples can be left in overnight; over decalcification is not a worry.
Packaging: Manufactured in the USA
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty