Would you like to log in again?
Formalin alternative PureFix™, Statlab
Supplier: StatLab
3180-5
76548-038EA 118.75 USD InStock
76548-038
Formalin alternative PureFix™, Statlab
Stains Microscopy and staining reagents
10% Non-buffered formalin.
  • Will not produce salt precipitates that may obstruct tissue processor lines
  • Preferred by some labs as a formalin alternative
Part of the StatLab line of prefilled bulk containers.
Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty