Would you like to log in again?
Xylene Substitute Citra-Clear™, Statlab
Supplier: StatLab
6400-1
76548-044EA 187.98 USD InStock
76548-044
Xylene Substitute Citra-Clear™, Statlab
Stains Microscopy and staining reagents
100% d-Limonene for processing and staining.
  • Mild citrus odor
  • Exceptionally high purity
  • Flash point 122°F
Part of the StatLab line of clearing agents.
Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty