Would you like to log in again?
Alcohol 95% Plus™, Statlab
Supplier: StatLab
7095-1
76548-054EA 27.06 USD InStock
76548-054
Alcohol 95% Plus™, Statlab
Stains Microscopy and staining reagents
A custom isopropyl and methanol blend that can be used as a substitute for traditional ethanol-based alcoholic blends.
  • Does not over-dehydrate small specimens
  • An excellent dehydrant for both small and large tissue specimens
  • May also be used in staining procedures
  • 100% Alcohol with 5 parts deionized water.
Part of the StatLab line of alcohol products that are ACS grade.
Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty