Would you like to log in again?
Zinc Formalin Buffered , Statlab
Supplier: StatLab
BFZ0230 BFZ0115 BFZ-1 BFZ0460 BFZ-5
76548-082CS 72.1 USD InStock
76548-082 76548-090 76548-086 76548-084 76548-088
Zinc Formalin Buffered , Statlab
Stains Microscopy and staining reagents
Ready-to-use Zinc Formalin solution.
  • Especially recommended for use in immunohistochemistry
  • Zinc formalin preserves more immunoreactivity than standard NBF
Part of the StatLab line of StatClick 10% NBF Prefills.
Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty