Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10% Modified Carson/Millonig

Supplier: StatLab
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

CM1115 CM1460 CM1507 CM1230
76548-092CS 69.93 USD InStock
76548-092 76548-096 76548-094 76548-098
Formalin 10% Modified Carson/Millonig
Formalin
A modified buffering formulation to preserve tissue.

  • Carson Millonig modified buffering formulation preserves tissue specimens
  • For both light and electron microscopy
  • Can be used in place of traditional 10% NBF

Part of the StatLab line of StatClick 10% NBF Prefills.

Manufactured in the USA


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty