Would you like to log in again?
PolarStat™ Embedding Medium, StatLab
Supplier: StatLab
CRYO-4
76548-100EA 49.65 USD InStock
76548-100
PolarStat™ Embedding Medium, StatLab
Stains Microscopy and staining reagents
An aqueous-based frozen section embedding medium designed to support tissue blocks in cryostat sectioning. Formulated to promote rapid freezing, enhanced sectioning and consistent results at a working temperature of -20C. 4 oz bottle.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty