Would you like to log in again?
Formalin Substitute GTF™, Statlab
Supplier: StatLab
FS-1
76548-114EA 19 USD InStock
76548-114
Formalin Substitute GTF™, Statlab
Stains Microscopy and staining reagents
A glyoxyl based, formalin-free fixative.
  • Excellent for conventional histology, special stains and immunohistochemistry
  • The formaldehyde-free solution provides the morphological detail of a standard aldehyde fixative while preserving high levels of immunoreactivity
Part of the StatLab line of formalin alternatives.
Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty