Would you like to log in again?
Millennia™ 1000 Adhesion Slides
Supplier: StatLab
Millennia™ 1000
M1000W/90 M1000G M1000Y M1000G/90 M1000P/90 M1000Y/90 M1000B M1000B/90 M1000W M1000P
76548-116BX 41.01 USD InStock
76548-116 76548-126 76548-124 76548-118 76548-128 76548-130 76548-134 76548-122 76548-132 76548-120
Millennia™ 1000 Adhesion Slides
Slides Adhesive Slides
Millennia™ 1000 slides feature a hydrophobic surface with strong adhesion properties.
  • Hydrophobic slide designed for IHC, special stains, frozen sections, and routine histology
  • Thermal transfer printer compatible
  • Available in both 45 degree clipped corners or 90 degree corners
Part of the StatLab Millennia™ Adhesion Microscope Slide family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty