Would you like to log in again?
Millennia™ 2000 Adhesion Slides
Supplier: StatLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Millennia™ 2000
M2000Y M2000G M2000W M2000B M2000P
76548-138BX 41.01 USD InStock
76548-138 76548-136 76548-140 76548-144 76548-142
Millennia™ 2000 Adhesion Slides
Slides Adhesive Slides
Millennia™ 2000 slides feature a hydrophilic surface with strong adhesion properties.
  • Hydrophilic slide designed for IHC, special stains, frozen sections, and routine histology
  • Thermal transfer printer compatible
  • Available with 45 degree clipped corners only
Part of the StatLab Millennia Adhesion Microscope Slide family. Manufactured in Japan.
Packaging: Slides are packaged 100 per box.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty