Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10%, neutral buffered

Supplier: StatLab
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

NB32480 NB0690 NB16240 NB08120
76548-168CS 137.64 USD InStock
76548-168 76548-170 76548-174 76548-172
Formalin 10%, neutral buffered
Formalin
Neutral buffered formalin prefill jars - large.

  • A full line of fixative options are conveniently prefilled and packed in StatLab’s proprietary HistoPak™ carrier for easy distribution
  • pH 7.0 ready-to-use buffered formalin

Part of the StatLab line of 10% NBF Prefills.

Manufactured in the USA


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty