Synonyms:
Methylaldehyde
, Formol
, Paraformaldehyde aqueous solution
, Formaldehyde
NB32480
NB0690
NB16240
NB08120
76548-168CS
137.64
USD
InStock
76548-168
76548-170
76548-174
76548-172
Formalin 10%, neutral buffered
Formalin
Neutral buffered formalin prefill jars - large.
- A full line of fixative options are conveniently prefilled and packed in StatLab’s proprietary HistoPak™ carrier for easy distribution
- pH 7.0 ready-to-use buffered formalin
Part of the StatLab line of 10% NBF Prefills.
Manufactured in the USA