Would you like to log in again?
Nail Prep™ Nail Softener, StatLab
Supplier: StatLab
NP-500
76548-176EA 139.57 USD InStock
76548-176
Nail Prep™ Nail Softener, StatLab
Stains Microscopy and staining reagents
Try this exclusive formulation designed for pre-processing nail preparation. Fast, efficient, and ready to use, with two concentrations available. Use pre-processing. The solution softens brittle nail clippings. Nail Prep™ makes specimens easier to cut and process, and helps to keep tissue from washing off the slide.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty