Would you like to log in again?
Prostate Biopsy Kit, StatLab
Supplier: StatLab
PBK0507 PBKNB12 PBKNB16 PBKNB6
76548-180EA 23.43 USD InStock
76548-180 76548-184 76548-182 76548-178
Prostate Biopsy Kit, StatLab
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
All StatLab prostate biopsy kits are pre-labeled with a location diagram supporting accurate collection. Kits contain prefilled StatClick™ vials with 10% neutral buffered formalin solution arranged in pre-labeled positions that correlate with the area of the prostate from which sample is taken, and stickers to affix to the prefill indicating the biopsy location. 6, 8, 12, or 16-vial biopsy kits are available. StatClick patented vials produce an audible “clicking” sound when the leak-proof seal has been properly engaged, assuring accurate closure and maintaining specimen integrity during transport.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty