Would you like to log in again?
Nail Prep™ Elite Nail Softener, StatLab
Supplier: StatLab
PH30-5
76548-190EA 220.26 USD InStock
76548-190
Nail Prep™ Elite Nail Softener, StatLab
Stains Microscopy and staining reagents
A faster Nail Prep! A higher concentration solution works quickly to soften brittle nail clippings. Use pre-processing. Results can be seen in as little as 15 minutes. 5 Gallon container.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty