Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High–Def® Acid Rinse, 3.5% Solution, StatLab

Supplier: StatLab
High–Def®
SL103
76548-212EA 56.62 USD InStock
76548-212
High–Def® Acid Rinse, 3.5% Solution, StatLab
Cleaning Agents Detergents and Surface Cleansers
For both Vintage and Reserve stain line use. A glacial acid based formulation. Provides unparalleled crisp nuclear chromatin detail and eliminates any undesired background staining when used with Reserve and Vintage Hematoxylin. Premixed, safe, and ready-to-use. High–Def® is also available in a low-concentration formulation for laboratories equipped with linear stainers.

Available in 1% and 3.5% Solutions.

Packaging: Manufactured in the USA

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty