Would you like to log in again?
StatFreeze™ Freezespray, 152a refrigerant, StatLab
Supplier: StatLab
SL202
76548-222CS 104.32 USD InStock
76548-222
StatFreeze™ Freezespray, 152a refrigerant, StatLab
Stains Microscopy and staining reagents
Ozone safe, does not contain CFC’s OR HCFC’s. Used to quickly cool paraffin blocks and fresh tissue in frozen sections. Sleek bottle is easy to hold and equipped with trigger nozzle and 5” tube to help direct spray.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty