Would you like to log in again?
Bluing Agent Reserve, Statlab
Supplier: StatLab
SL203
76548-224EA 54.59 USD InStock
76548-224
Bluing Agent Reserve, Statlab
Stains Microscopy and staining reagents
Converts the soluble red component of hematoxylin into an insoluble blue.
  • A buffered alkaline rinse with a mild pH (8.0)
  • Ensures optimal staining and proper cellular contrast
  • Bluing reagent is premixed and ready-to-use.
Part of the StatLab Reserve stain line.
Manufactured in USA.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty