Would you like to log in again?
Tissue Marking Dyes, StatLab
Supplier: StatLab
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SL669 SL662VT-2 SL662GR-2 SL662BL-2 SL662YL-2 SL662BK-2 SL662OR-2 SL662RD-2
76548-248EA 39.77 USD InStock
76548-248 76548-246 76548-240 76548-250 76548-244 76548-254 76548-242 76548-252
Tissue Marking Dyes, StatLab
Stains Histology Stains
Permanent tissue marking dyes. Seven colors formulated to define margins without bleeding or fading. Durable, bright dyes hold permanently through processing and staining procedures. Unique lids may either be completely removed or conveniently flipped open to prevent drying.
Set includes one tissue marking dye rack and a set of 2 oz dyes in each of 7 colors.
Packaging: Manufactured in the USA
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty