Would you like to log in again?
TissueGuard™ Specimen Processing Gel, StatLab
Supplier: StatLab
TG12
76548-294BX 407.42 USD InStock
76548-294
TissueGuard™ Specimen Processing Gel, StatLab
Stains Microscopy and staining reagents
tabilize fragile specimens during processing by preparing with TissueGuard processing gel. Tissue Guard encapsulates and retains specimens during histological processing while allow chemical reagents to penetrate the specimen. Appropriate for cytology and histology applications, TissueGuard is an alternate to other agarose stabilizers.
  • 12 tubes per kit
  • 10 mL per tube
Cytology Protect fine needle aspirates, urine specimens, non-gyn specimens, and any other specimens utilizing cell block preparation. Product can be applied in either centrifuge tube, or in conjunction with BioMesh tissue cassettes, or using tissue paper. After application, specimen is ready for processing within minutes, or sooner if aided with a cooling block. Histology Protect samples including needle biopsies, nerves, small arteries, aggregates, lymph nodes and the smallest of tissue fragments. Product can be applied to samples in conjunction with BioMesh cassettes, or using filter paper.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty