Would you like to log in again?
Urofix Fixative for Prostate Biopsies, StatLab
Supplier: StatLab
UF0507 UF0230 UF0460
76548-302CS 69.18 USD InStock
76548-302 76548-306 76548-304
Urofix Fixative for Prostate Biopsies, StatLab
Stains Microscopy and staining reagents
An excellent fixative for prostate biopsies. Preserves nuclear detail extremely well, and penetrates tissue quicker, thereby shortening fixation times.
Packaging: Manufactured in the USA
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty