Would you like to log in again?
Scissors, Iris, Curved, Mortech
Supplier: Mortech
AA129 AA029
76548-458EA 98.41 USD InStock
76548-458 76548-364
Scissors, Iris, Curved, Mortech
Scissors
Ideal for small, precise, post-mortem dissection.
  • Stainless steel
  • Curve cutting edge
  • Sharp points
Available in standard or premium grade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty