Would you like to log in again?
Scissors, Tungsten Carbide, Metzenbaum, Mortech
Supplier: Mortech
AA411 AA407 AA408 AA412 AA413 AA415 AA410 AA409 AA414
76548-534EA 317.47 USD InStock
76548-534 76548-544 76548-532 76548-542 76548-538 76548-548 76548-536 76548-546 76548-540
Scissors, Tungsten Carbide, Metzenbaum, Mortech
Scissors
The unique tungsten carbide cutting edge provides effortless, yet accurate cuts through the toughest of tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex post-mortem dissection cases.
  • Tungsten carbide
  • Narrow, blunt tips
Available with straight or curved edges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty