Would you like to log in again?
Scissors, SuperCut Mayo, Mortech
Supplier: Mortech
AA522 AA538 AA521 AA520 AA519 AA537
76548-578EA 349.68 USD InStock
76548-578 76548-598 76548-576 76548-580 76548-574 76548-596
Scissors, SuperCut Mayo, Mortech
Scissors
SuperCut scissors are tempered and for a longer, sharper life over ordinary steel. The SuperCut blade has a specially designed razor sharp edge. The straight version can cut into surface area tissue while the curved version can cut into heavier or deeper tissue. They also feature rounded and polished finger rings.
  • Stainless steel
  • Slightly rounded tips
Available with straight or curved edges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty